Eisco Premium Bunsen Burner with Flame Stabilizer
Supplier: Wards
CH0094LP CH0094NG
470343-838EA 53.55 CAD
470343-838 470344-866
Eisco Premium Bunsen Burner with Flame Stabilizer
Heaters Gas Burners
With both air mix adjustment and gas flow adjustment.
  • Suitable for use with LPG (liquid petroleum gas) or butane gas
  • Serrated inlet arm accepts 6 to 8 mm tubing
  • Aluminum mixing tube is mounted on a circular, die casted zinc alloy base
  • Equipped with a flame stabilizing burner tip and air ports with collar to adjust the flow of air
  • Gas flow is controlled by needle valve fitted above the base, permitting easy flame adjustment
The flame stabilizer makes this burner safer than standard Bunsen burners. Features nickel-plated die casted zinc alloy base, aluminum mixing tube, flame stabilizing brass burner tip and adjustable air ports. Air supply is controlled by a nickel plated brass sleeve covering ports at the lower end of the mixing tube, and gas flow is controlled by needle valve fitted above the base for easy flame adjustment. Burner measures 6.5" tall, and the base measures 3" in Ø.