Eisco Glass Aspirator Bottle with Ribbed Outlet
Supplier: Wards
CH0060B CH0060A CH0060C
470344-260EA 42.8 CAD
470344-260 470344-398 470344-912
Eisco Glass Aspirator Bottle with Ribbed Outlet
Bottles Aspirators
Ungraduated glass aspirator bottles have a ribbed sidearm outlet located at the base of the bottle for attaching tubing and an open, beaded neck.
  • Made of high-quality, heavy-duty, borosilicate 3.3 glass
  • Superior durability and chemical resistance
  • Can withstand high temperatures and thermal shocks
  • Integral ribbed outlet at base of bottle for attaching tubing
Aspirator bottles are autoclavable, and offers superior chemical durability and a low temperature gradient. They can withstand high temperatures and thermal shocks.
Eisco borosilicate glass hasMore Product Information