Silicone Stoppers, United Scientific Supplies
Supplier: United Scientific Supplies
UNSST0 UNSST00 UNSST1 UNSST2 UNSST3 UNSST4 UNSST5 UNSST5A UNSST6 UNSST6A UNSST7 UNSST7A UNSST8
470340-262EA 10.98 CAD
470340-262 470340-104 470339-738 470338-900 470339-230 470340-132 470339-790 470339-228 470338-876 470340-000 470339-174 470339-106 470339-748
Silicone Stoppers, United Scientific Supplies
Stoppers Silicone Stoppers
Chemical resistant solid silicone stoppers.
  • Sturdy non reactive silicon
Silicone stoppers are particularly useful due to their inert and nonreactive nature. It will not impair the growth of tissue cultures and resistant to most chemical acids and solvents. Stoppers can be steam sterilized at 15 lbs. pressure. Broad working temperature range of –62 to +232 °C (–80 to +450 °F).
Durometer hardness ~50.