United Scientific™ Magnetic Stir Bars, Cross-Shaped, United Scientific Supplies
Supplier: United Scientific Supplies
MXB38 MXB42
470339-832EA 16.2 CAD
470339-832 470339-722
United Scientific™ Magnetic Stir Bars, Cross-Shaped, United Scientific Supplies
Stirrers Magnetic Stirring Bars
Cross-shaped magnetic stirring bars.
  • PTFE coated
  • High quality magnets
Magnetic stir bars are constructed of high quality magnets coated with PTFE. Cross-shaped stirring bars increase turbulence and improve stirring action.