Racks For Microcentrifuge Tubes, PC, United Scientific Supplies
Supplier: United Scientific Supplies
77705 77705-A
470339-282EA 74.5 CAD
470339-282 470340-150
Racks For Microcentrifuge Tubes, PC, United Scientific Supplies
Racks Tube Racks
Polycarbonate racks for 1.5 ml microcentrifuge tubes.
  • Autoclavable
  • Include alphanumeric grid for easy identification of tubes
Unique design allows racks to be securely stacked on top of each other.