Cryo Cube Boxes, PP, United Scientific Supplies
Supplier: United Scientific Supplies
66501 66502
470338-952EA 55.45 CAD
470338-952 470339-822
Cryo Cube Boxes, PP, United Scientific Supplies
Boxes Cryoboxes
Polypropylene cryo boxes with alpha-numeric grid.
  • Autoclavable
  • Temperatures range: –90 °C to +121 °C.
Manufactured of polypropylene, these autoclavable boxes are stackable for sample storage in 1.5 to 2.0 ml microtubes and 1.0 to 2.0 ml cryogenic vials. Boxes feature a hinged lid connected to the base and a locking system to ensure secure closure. A molded alphanumeric grid allows for easy identification of samples. Dimensions of cryo boxes are designed to be compatible with vertical, horizontal, and drawers style freezer racks. Each pack includesMore Product Information