Buchner Funnel, Porcelain, United Scientific Supplies
Supplier: United Scientific Supplies
JBF600 JBF800 JBF003 JBF035 JBF100 JBF1150 JBF200 JBF2000 JBF350 JBF4800
470339-322EA 59.4 CAD
470339-322 470339-550 470338-924 470339-438 470339-864 470340-030 470338-964 470339-902 470339-236 470339-186
Buchner Funnel, Porcelain, United Scientific Supplies
Funnels Filtration Funnels Büchner Funnels
Buchner Funnels for filtering hot or cold solutions.
  • Resistant to chemical action
  • Glazed on inner and outer surfaces, excluding rims; no softening occurs up to 1000 °C
  • Thermal shock resistant
High-quality porcelain Buchner funnel with 3ml capacity and fixed, perforated plates. Funnel is vibrant white and glazed on the inner and outer surfaces (except the rims), designed to withstand softening at up to 1000 °C. Highly resistant to nearly all acids and alkalis, excluding hydrofluoric acid.