Spray Bottles, PET, United Scientific Supplies
Supplier: United Scientific Supplies
UNSBOT16OZPK8 UNSBOT16OZBULK UNSBOT8OZ UNSBOT80ZPK12
470339-364PK 509.85 CAD
470340-038 470339-412 470339-364 470339-938
Spray Bottles, PET, United Scientific Supplies
Bottles Spray Bottles
Clear PET spray pump bottle.
  • PET bottle
  • Spray nozzle
  • Chemical-resistant
Spray bottles are made from clear polyethylene terephthalate (PET) and include a white spray nozzle with clear cap to keep nozzle clean and leak-proof. PET is lightweight, chemically-resistant, and provides excellent durability.