Spatula with Spoon, Porcelain, United Scientific Supplies
Supplier: United Scientific Supplies
JSS060 JSS010 JSS001 JSS001 JSS003
470339-202EA 42.58 CAD
470339-202 470340-236 470340-186 470339-662 470339-848
Spatula with Spoon, Porcelain, United Scientific Supplies
Spatulas
Chemical-porcelain spatulas feature a spatula on one end and a spoon on the other.
  • Glazed porcelain laboratory spatula with one flat end and one spoon end
  • Autoclavable
This glazed porcelain laboratory spatula is designed with one flat end and one spoon end.
Order Now
SPECIFICATIONS
Environmentally Preferable