Porcelain Crucible, Gooch Filter, with Cover, United Scientific Supplies
Supplier: United Scientific Supplies
JGC015 JGC030 JGC050 JGC140
470339-568EA 64.54 CAD
470339-568 470339-466 470339-312 470339-382
Porcelain Crucible, Gooch Filter, with Cover, United Scientific Supplies
Crucibles
Crucibles for determination of total suspended solids.
  • Autoclavable
  • Glazed inside and out, except for outside bottom and rim
  • Maximum use temperature of 1150 °C (2102 °F)
  • Covers included
Gooch crucibles are ideal for the determination of total suspended solids. Excellent resistance to acids and alkalies, not including hydrofluoric acid. Autoclavable and glazed inside and out, except for outside bottom and rim. Designed to withstand temperatures up to 1150 °C; to prevent thermal stress cracks, temperature change rate should no exceed 200 °C per hour.