High Alumina Crucibles, Tall Form, United Scientific Supplies
Supplier: United Scientific Supplies
JAH010 JAH020 JAH050 JAH100
470340-142EA 50.64 CAD
470340-142 470339-400 470339-208 470339-740
High Alumina Crucibles, Tall Form, United Scientific Supplies
Crucibles
Tall form high alumina crucibles.
  • Made from 99.7% pure aluminum oxide for maximum use temperature of 1750 °C (3182 °F), Temperature change rate should not exceed
  • 150 °C (302 °F) per hour
  • Recommended for use with refractory metals, suitable for glass melting
This cylindrical-form crucible with 10 ml capacity is crafted from 99.7% pure aluminum oxide, ideal for high-temperature work demanding contamination-free results. Maximum use temperature is 1750 °C (3182 °F); temperature change rate should not exceed 150 °C (302 °F) per hour.
High-aluminaMore Product Information