Tris(2-chloroethyl)phosphate ≥93.0% (by GC)

Supplier: TCI America
Danger

T3447-100G T3447-25G
TCT3447-100GEA 124.82 CAD
TCT3447-100G TCT3447-25G
Tris(2-chloroethyl)phosphate ≥93.0% (by GC)
Tris(2-chloroethyl) phosphate
Formula: C₆H₁₂Cl₃O₄P
Boiling Pt: 146°C
Flash Pt: 222°C
MDL Number: MFCD00000967
CAS Number: 115-96-8
EINECS: 204-118-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR