2,2,6,6-Tetramethyl-4-(2-propynyloxy)piperidine 1-oxyl ≥98.0% (by GC, titration analysis) free radical

Supplier: TCI America
Warning

T3169-1G T3169-5G
TCT3169-1GEA 184.86 CAD
TCT3169-1G TCT3169-5G
2,2,6,6-Tetramethyl-4-(2-propynyloxy)piperidine 1-oxyl ≥98.0% (by GC, titration analysis) free radical
2,2,6,6-Tetramethyl-4-(2-propynyloxy)piperidine 1-oxyl
Formula: C₁₂H₂₀NO₂
MDL Number: MFCD01126982
CAS Number: 147045-24-7

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 98.0 %
Melting point 65.0 to 69.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR