Sodium tetrafluoroborate ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Sodium fluoroborate

O0494-25G O0494-500G
TCO0494-25GEA 55.73 CAD
TCO0494-25G TCO0494-500G
Sodium tetrafluoroborate ≥95.0% (by titrimetric analysis)
Sodium tetrafluoroborate
Formula: BF₄Na
MW: 109.79 g/mol
Melting Pt: 384°C
MDL Number: MFCD00003515
CAS Number: 13755-29-8
EINECS: 237-340-6
Merck Index: 14,08617

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR