Ethoxy(pentafluoro)cyclotriphosphazene ≥98.0% (by GC)
Supplier: TCI America
|
Danger
|
Synonyms:
2-Ethoxy-2,4,4,6,6-pentafluoro-1,3,5,2,4,6-triazatriphosphorine
E1140-1G
E1140-5G
TCE1140-1GEA
175.38
CAD
TCE1140-1G
TCE1140-5G
Ethoxy(pentafluoro)cyclotriphosphazene ≥98.0% (by GC)
Ethoxy(pentafluoro)cyclotriphosphazene
Formula:
C₂H₅F₅N₃OP₃ MW: 275.00 g/mol Boiling Pt: 42°C Storage Temperature: Refrigerator |
MDL Number:
MFCD28386107 CAS Number: 33027-66-6 UN: 1760 ADR: 8 III |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...