Bis(2,2,6,6-tetramethyl-4-piperidyl-1-oxyl) Sebacate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
Bis(2,2,6,6-tetramethyl-4-piperidyl-1-oxyl) Decanedioate, Decanedioic acid bis(2,2,6,6-tetramethyl-4-piperidyl-1-oxyl) ester, Sebacic acid bis(2,2,6,6-tetramethyl-4-piperidyl-1-oxyl) ester
B5642-1G
B5642-5G
TCB5642-1GEA
175.38
CAD
TCB5642-1G
TCB5642-5G
Bis(2,2,6,6-tetramethyl-4-piperidyl-1-oxyl) Sebacate ≥98.0% (by titrimetric analysis)
Bis(2,2,6,6-tetramethyl-4-piperidyl-1-oxyl) Sebacate
Formula:
C₂₈H₅₀N₂O₆ MW: 510.72 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12756000 CAS Number: 2516-92-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Iodometric Titration) | min. 98.0 % |
Melting point | 99.0 to 103.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...