Millennia™ Command Adhesion Slides, StatLab Medical Products
Supplier: StatLab
MCOMM/A/90 MCOMM/G/90 MCOMM/L/90 MCOMM/P/90 MCOMM/T/90 MCOMM/Y/90
76553-854BX 65.32 CAD
76553-854 76553-842 76553-850 76553-844 76553-852 76553-846 76553-848
Millennia™ Command Adhesion Slides, StatLab Medical Products
Slides Microscope Slides
A universal adhesion slides for H&E, IHC and special stains.
  • Hybrid slides suitable for IHC, H&E, and special staining
  • Optimized coating for strong adhesion to reduce tissue loss and provide consistency of staining
  • Exhibits minimal background staining during all types of staining
  • Hydrophilic, 90° corners
  • Compatible with thermal transfer printers
Part of the StatLab Millennia adhesion microscope slides family. Manufactured in Japan.
Packaging: Slides are packaged 100 per box.