Sequential Sterilization Wraps, Halyard
Supplier: O&M HALYARD, INC
10715 10718 10720 10730 10736 10740 10745 10748 68015 68018 68036 68054 68072 10830 10840 10845 10854 10872 68245 68254 68118 68124 68130 68136 68154 68172 68636 68640 68645 68648 68654 68672
76557-812CS 272.3 CAD
76557-812 76557-814 76557-816 76557-818 76557-820 76557-822 76557-824 76557-826 76557-828 76557-830 76557-832 76557-834 76557-836 76557-838 76557-840 76557-842 76557-844 76557-846 76557-848 76557-850 76557-852 76557-854 76557-856 76557-858 76557-860 76557-862 76557-864 76557-866 76557-868 76557-870 76557-872 76557-874
Sequential Sterilization Wraps, Halyard
Sterilization Wraps
The choice of sterile processing professionals, HALYARD sterilization wraps, made of strong reliable SMS non-woven fabric, have been helping keep sterilized instruments and supply sets ready for use at a moment’s notice. Ideal for use in individual medical and dental practices as well as major medical and surgery centers.
  • Industry standard for sterile processing teams around the world
  • Tear-, flame-, lint- and abrasion-resistant
  • Pre-vacuum steam sterilant penetration
  • Maintenance of package integrity (MPI) to meet FDA requirements
  • Ethylene oxide (EO)
More Product Information