Transfer Isolators, 2 Gloves, Rheo Flexibles (Division of Rheo Engineering)
Supplier: RHEO ENGINEERING LLC
4001788 4001789 4001790
76557-666EA 3433.8 CAD
76557-666 76557-756 76557-652
Transfer Isolators, 2 Gloves, Rheo Flexibles (Division of Rheo Engineering)
Glove Boxes and Glove Bags
The Rheo transfer isolator is a single-use isolator that provides high containment operator protection during the handling of potentially hazardous powders. This quick deployment solution can be installed quickly and can be disposed of when finished, eliminating the cleaning costs associated with traditional stainless steel isolators.
  • Constructed from RheoFlex 200 series film polyurethane material
  • FDA compliant
  • 21 CFR 177.1680 compliant
  • Clear and matte finish
  • Anti-static film
  • BSE/TSE compliant
The bungee strap system and the general flexibility ofMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable