RheoCrimps, Rheo Flexibles (Division of Rheo Engineering)
Supplier: RHEO ENGINEERING LLC
4000466 4001354
76557-642EA 4860.14 CAD
76557-642 76557-658
RheoCrimps, Rheo Flexibles (Division of Rheo Engineering)
Crimpers, Cappers and Decappers
The RheoCrimp is an easy to use cut and crimp separation technology. These crimps offer a wide opening range for capturing bulky liner material. The RheoCrimp kit includes the tools needed for this crimp and cut process.
  • Wide-range crimp capability
  • Dual-tie design
  • Easy alignment cut-notch for cutting operation
  • Grip-tight locking rib to ensure integrity
The wrap-around design of a tie-wrap allows for the operator to easily envelop a wide range of material, with little twisting and bunching.
Ordering information: Kit includes Cable -Tie tensioner, 50 RheoCrimps, 50 zip ties, roll of GMP tape, and cutting shears for cutting between Cable-Ties. Each RheoCrimp pack includes 50 RheoCrimps.
Packaging: All components packaged in tidy carrying case.