Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
M00652 M00653 M00654 M00655 M00656 M00657 M00658 M00659 M00660 M00661 M00662 M00663 M00664 M00665 M00666 M00667 M00668 M00669 M41210 M41212 M41215 M42010 M42012 M42015
76510-572EA 165.65 CAD
76510-572 76510-574 76510-576 76510-578 76510-580 76510-582 76510-584 76510-586 76510-588 76510-590 76510-592 76510-594 76510-596 76510-598 76510-600 76510-602 76510-604 76510-606 76510-656 76510-658 76510-660 76510-662 76510-664 76510-666
Bis-Tris Mini Protein Gels, GenScript®
Electrophoresis Gels Precast Gels
GenScript's Bis-Tris gels are high-performing polyacrylamide gels designed to separate a wide range of protein sizes by electrophoresis. The gels are cast in a neutral pH buffer that minimizes polyacrylamide hydrolysis and increases gel stability. Furthermore, the neutral pH which minimizes protein modification when compared to Tris-glycine gels.
  • Large well volume
  • High resoluton and reproducibility
  • Long shelf life
  • Cost effective
  • Compatible cassette design
SurePAGE™ gels are premium, high resolution with enhance reproducibility gels, while the ExpressPlus are good quality and cost-effective gels.
Proteins are separated on SurePAGE using MOPS or MES running buffer. We recommend using MOPS buffer for protein sizes that are >30 kD and MES for proteins that are <30 kD.
We recommend using 4×LDS sample buffer (M00676) as the sample loading buffer.