Innovating Science® Forensic Chemistry of Unknown Substances
Supplier: Ward's Science
IS1507 IS9006
470329-584EA 69.9 CAD
470329-584 470343-400
Innovating Science® Forensic Chemistry of Unknown Substances
Educational Classroom Kits and Activities
Learn about how investigators identify unknown substances to help solve crimes.
  • Students will use observation skills, senses, and chemical tests on a series of known substances as well as two unknown substances to attempt to identify the mystery substances
  • Aligned to NGSS Practices
  • Performance expectations: MS-PS1-2
  • Disciplinary core ideas: PS1.A, PS1.B
  • Cross cutting concepts include Patterns
  • Engineering practices include analyzing and interpreting data
Often times, when collecting evidence at a crime scene, investigators may recover substances they are unable to identify in the field. Along with evidence such as fingerprints, hair, fibers, etc., there may be traces of unknown chemicals or powders left behind by the perpetrator or perpetrators. Evidence of this nature is sent to the crime lab for identification.
In this experiment, you will use your observation skills, senses, and chemical tests on a series of known substances as well as two unknown substances. You will then attempt to identify the mystery substances based on your observations and recorded data.
Kit contains enough material for 15 groups. Teacher's Manual and Student Study Guide copymasters are included.
Delivery information: Instructor's manual and SDS included.
Caution: This product can expose you to chemicals including Respirable crystalline silica, which is known to the State of California to cause cancer. For more information go to http://www.P65Warnings.ca.gov.