Verline™ Lead-Free Beige Autoclave Indicator Tapes, Propper Manufacturing Company
Supplier: Propper
26810500 26810600
76557-782CS 466.1 CAD
76557-782 76557-786 76557-783 76557-787
Verline™ Lead-Free Beige Autoclave Indicator Tapes, Propper Manufacturing Company
Sterilisation Indicators
Verline™ lead free autoclave steam indicator tape is pressure-sensitive sterilization tape that is used to distinguish between processed and unprocessed items.
  • Distinguish between processed and unprocessed sterilization packages
  • Strong crepe-embossed, latex-free tape
  • Distinct green to dark brown/black color change after exposure to steam
  • For use in securing wrapped surgical trays before, during, and after sterilization
The lead free indicator ink placed in green stripes turn dark brown/black after exposure to a steam sterilization cycle