Thermal Conductivity Detector (TCD), Agilent Technologies
Supplier: Agilent
G3432-60220 G3432-60220
CAG3432-60220EA 4888.69 CAD
AGG3432-60220 CAG3432-60220
Thermal Conductivity Detector (TCD), Agilent Technologies
Chromatography Detectors
Detector responds to all compounds and compares the thermal conductivities of two gas flows using a single-filament, single-column design.
  • Designed, tested and manufactured by Agilent engineers to work perfectly with our instruments Superior performance
The thermal conductivity detector responds to all compounds, excluding carrier gas. The TCD compares the thermal conductivities of two gas flows – pure carrier gas (also called the reference gas) and carrier gas plus sample components (also called column effluent), using a single-filament, single-column design.