Cell Spacers and Inserts for UV-Vis and UV-Vis-NIR, Agilent Technologies
Supplier: Agilent
5061-3390 5061-3388 5061-3390 5061-3389 5061-3389 5061-3388
CA5061-3390EA 184.44 CAD
AG5061-3389 AG5061-3388 CA5061-3390 CA5061-3388 AG5061-3390 CAAG5061-3389
Cell Spacers and Inserts for UV-Vis and UV-Vis-NIR, Agilent Technologies
Cuvettes Spectrophotometer Cuvettes
Spacers allow 1, 2, and 5 mm path length cells (with less than 12.5 mm depth) to be securely mounted in a standard cell holder (for 12.5×12.5 mm cells).
  • Required when there is no path length adjustment facility in the cell holder
  • Quartz block inserts can also be used instead of spacers - they provide more flexibility as each insert provides two different pathlengths selected by rotating through 90 degrees
Spacers or inserts are required for short path length cells that have less than 12.5 mm depth, to ensure secure mounting in a standard cell holder. Agilent offers black anodized aluminum spacers and quartz block inserts for use with 1, 2, and 5 mm path length cells.