µSIL-WAX Capillaries with Windows, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
196-7203 197-7202
CAAG196-7203PK 638.37 CAD
CAAG196-7203 CAAG197-7202
µSIL-WAX Capillaries with Windows, Agilent Technologies
Electrophoresis Systems Capillary Electrophoresis Systems
µSIL-WAX capillaries with windows, featuring a modified, polyethylene oxide, hydrophilic coating.
  • Ideal for CE/MS
  • Ideal for protein and peptide separations from pH 2 to 5
µSIL-WAX capillaries feature a modified, polyethylene oxide, hydrophilic coating. The coating effectively masks active silanol sites. The highly stable coating and near-zero EOF of µSIL-WAX makes the capillary ideal for CE-MS, and protein and peptide separations.