PROPOR SG MURUS Capsules, Parker Bioscience
Supplier: PARKER HANNIFIN TE
ZLSGK-010BB-PNL ZLSG1-010BB-PNL ZLSG2-010BB-PNL ZLSG3-010BB-PNL ZLSGK-020BB-PNL ZLSG1-020BB-PNL ZLSG2-020BB-PNL ZLSG3-020BB-PNL ZLSGK-045BB-PNL ZLSG1-045BB-PNL ZLSG2-045BB-PNL ZLSG3-045BB-PNL ZLSGK-010BB-PSL ZLSG1-010BB-PSL ZLSG2-010BB-PSL ZLSG3-010BB-PSL ZLSGK-020BB-PSL ZLSG1-020BB-PSL ZLSG2-020BB-PSL ZLSG3-020BB-PSL ZLSGK-045BB-PSL ZLSG1-045BB-PSL ZLSG2-045BB-PSL ZLSG3-045BB-PSL ZLSG1-020AA-PNL-S ZLSG1-020AA-PSL-S ZLSG1-020HH-PNL-S ZLSG1-020HH-PSL-S ZLSG2-020AA-PNL-S ZLSG2-020AA-PSL-S ZLSG2-020HH-PNL-S ZLSG2-020HH-PSL-S ZLSG3-020AA-PNL-S ZLSG3-020AA-PSL-S ZLSG3-020HH-PNL-S ZLSG3-020HH-PSL-S ZLSGK-020AA-PNL-S ZLSGK-020AA-PSL-S ZLSGK-020HH-PNL-S ZLSGK-020HH-PSL-S
76552-118EA 518.48 CAD
76552-118 76552-120 76552-122 76552-124 76552-126 76552-128 76552-130 76552-132 76552-134 76552-136 76552-138 76552-140 76552-142 76552-144 76552-146 76552-148 76552-150 76552-152 76552-154 76552-156 76552-158 76552-160 76552-162 76552-164 77676-166 77676-174 77676-164 77676-162 77676-180 77676-172 77676-170 77676-178 77676-152 77676-156 77676-182 77676-168 77676-154 77676-158 77676-160 77676-176
PROPOR SG MURUS Capsules, Parker Bioscience
Filters Membrane Filter Capsules
PROPOR SG sterilizing grade filters feature a microbially retentive polyethersulfone membrane for fast, reliable and cost-effective sterile filtration of pharmaceutical fluids.
  • Up to 3.5 times higher flow rates than competitive sterilizing grade filters
  • Fully validated and integrity testable membrane for assurance of sterility
  • Low binding for minimal product loss
  • Single-use capsule format available with Pre-sterile version by Gamma-irradiation
The asymmetric pore structure and high voids volume of the PROPOR SG membrane allow high throughputs and exceptionally high flow rates compared with competitive PES and alternative membranes. Low protein and preservative binding properties minimize product loss due to adsorption.
PROPOR SG filters are optimized for pharmaceutical processing. They have low extractable levels and broad chemical compatibility across the full pH range including organic solvents.