Fittings for Thermo/Dionex LC Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8002-0802 8002-0921
CAAG8002-0802EA 63.03 CAD
CAAG8002-0802 CAAG8002-0921
Fittings for Thermo/Dionex LC Systems, Agilent Technologies
Fittings and Connectors
Achieve leak-free connections on selected Thermo Fisher Scientific HPLC systems.
  • Robust design for long life and smooth, leak-free operation
  • Stainless steel nuts and ferrules for high-pressure systems
  • Polymer fittings for biocompatible applications
Agilent fittings for selected Thermo Fisher Scientific HPLC systems give you added confidence and choice by helping to ensure leak-free connections and prevent loss of peak shape and resolution.
A variety of recommended fitting styles for columns, valves, and unions are available.