Cleanup Cartridges, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
GC210-4 GC250-4 GKI-4025 GKI-4756
CAAGGC210-4PK 1914.29 CAD
CAAGGC210-4 CAAGGC250-4 CAAGGKI-4025 CAAGGKI-4756
Cleanup Cartridges, Agilent Technologies
Chromatography Columns
Remove contaminants with fast and simple glycan cleanup options for a variety of workflows
  • Cleanup up of glycans is often performed before labeling to remove contaminating proteins, as well as after labeling to improve data quality and extend column life.
  • Cleanup cartrides are intended to complete a user-developed traditional N-glycan prep method.
Remove contaminants from glycan samples with simple protocols. With cartridges available for protein removal, desalting, excess dye removal, and other applications, all areas of glycan sample purification are covered. Whether your traditional workflow demands high-throughput or a few samples at a time, there is a cleanup option for you.