Bond Elut SAX Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
7540802C A3960850
CAAG7540802CEA 909.16 CAD
CAAG7540802C CAAGA3960850
Bond Elut SAX Plates, Agilent Technologies
Microplates Solid Phase Extraction Plates (SPE)
Bond Elut SAX is a strong anion-exchange sorbent well suited to compounds such as carboxylic acids.
  • Retains compounds that elute from weak anion-exchange sorbents
  • Selectivity can be user-modified for increased flexibility
  • Minimal nonpolar interactions
Bond Elut SAX is a strong anion-exchange sorbent ideally suited for the extraction of compounds, such as carboxylic acids, which may not retain effectively on weak anion-exchange sorbents.