Bond Elut Plexa PAX Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5982-0601 A3967010 A3967030 A4967010 A4967030
CAAG5982-0601EA 843.13 CAD
CAAG5982-0601 CAAGA3967010 CAAGA3967030 CAAGA4967010 CAAGA4967030
Bond Elut Plexa PAX Plates, Agilent Technologies
Microplates Solid Phase Extraction Plates (SPE)
Bond Elut Plexa PAX nonpolar/anion-exchange SPE is suitable for polar and nonpolar acidic analytes.
  • Unique phenylboronic acid sorbent
  • High specificity for cis-diol compounds
  • Suitable for a broad range of biomolecule applications
Bond Elut Plexa PAX lifts the performance standard in analyte cleanup and reproducibility for polar and nonpolar acidic analytes. Plexa PAX particles are functionalized using a proprietary process, allowing anion-exchange loadings to be controlled with high reproducibility. This gives more robust performance across yourMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable