Bond Elut C18 EWP Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
7540103C 7542601C
CAAG7540103CEA 798.67 CAD
CAAG7540103C CAAG7542601C
Bond Elut C18 EWP Plates, Agilent Technologies
Microplates Solid Phase Extraction Plates (SPE)
Bond Elut C18 EWP features wide pores, making it ideal for efficient extraction of macromolecules.
  • No exclusion of large molecules
  • Good for desalting proteins
  • Successful separation of proteins, peptides, or nucleotides
Bond Elut EWP is based upon standard particle size silica but with 500 Å pores to allow more efficient extraction of large molecules (>15,000 MW), which are typically excluded from standard porosity silica phases. Typical matrices include aqueous samples and biological fluids.