Dissolution Sinkers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
12-3000 12-3070
CAAG12-3000EA 494.11 CAD
CAAG12-3000 CAAG12-3070
Dissolution Sinkers, Agilent Technologies
Dissolution Apparatus
A variety of dissolution sinkers are available depending on your dosage form.
  • Capsule wire, weights, and sinker baskets are used to retain a dosage form at the bottom of the vessel for testing when the dosage form is buoyant
  • A variety of options are available
  • The flexible capsule wire meets USP requirements and has a 0.032" diameter
  • Sinker baskets are offered in a variety of mesh sizes and made of stainless steel with press-on caps, using O-rings to seal the basket
  • Japanese Pharmacopoeia (JP) sinker baskets are available as an alternative
  • Three-prong
More Product Information