Dissolution Evaporation Covers, Agilent Technologies
Supplier: Agilent
12-6327 12-6328 12-6315 12-6315 12-6311 12-6330 27-1500 12-6328 12-6305 12-6300 12-6311 12-6310 12-6305 12-6330 27-1510 12-6327 12-6329 12-6300 12-6320 12-6329 12-6320 12-6310 27-1500 27-1510
CAAG12-6328EA 644.78 CAD
CAAG12-6328 AG12-6329 CAAG12-6305 CAAG12-6327 AG12-6327 CAAG12-6315 AG12-6305 AG12-6328 CAAG12-6329 CAAG12-6320 AG12-6310 AG12-6311 CAAG12-6330 AG12-6300 AG12-6330 AG12-6320 AG12-6315 CAAG12-6311 CAAG12-6300 CAAG12-6310 AG27-1500 CAAG27-1510 AG27-1510 CAAG27-1500
Dissolution Evaporation Covers, Agilent Technologies
Dissolution Apparatus
A variety of evaporation covers are available to fit your apparatus and vessels.
  • The evaporation covers automatically travel with the apparatus driveunit using either the alignment posts or DDM assemblies
  • The standard evaporation cover on the 708-DS is designed to allow less than 1% evaporative loss over a 24-hour period under normal operating conditions
  • Agilent also offers other low loss covers that can reduce evaporative loss even further
A variety of evaporation covers are available to fit your apparatus and vessels. Evaporation plugs are used to minimize evaporation by sealing unused ports in the evaporation covers.