Syringes for Dionex HPLC Autosamplers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8002-0405 8002-0407
CAAG8002-0405EA 357.65 CAD
CAAG8002-0405 CAAG8002-0407
Syringes for Dionex HPLC Autosamplers, Agilent Technologies
Syringes Syringes for Chromatography
Agilent syringes for Dionex autosamplers ensure accurate and effective sampling.
  • Accuracy to within ±1% of nominal volume with a precision of 1% at 80% of the total volume
  • A chemically inert fluid path of stainless steel, borosilicate Type I glass, or PTFE
  • Robust design for long lifetime
  • Chem and removable syringe terminations are available
The Agilent HPLC syringe portfolio for Dionex autosamplers contains a broad selection of syringe styles and volumes to provide what you need for accurate and effective sampling. From autosampler syringes in bothMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable