Capillaries for Thermo/Dionex LC Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8002-0803 8002-0818 8002-0819 8002-0820 8002-0821 8002-0822 8002-0823 8002-0824 8002-0825 8002-0826
CAAG8002-0803EA 178.32 CAD
CAAG8002-0803 CAAG8002-0808 CAAG8002-0818 CAAG8002-0819 CAAG8002-0820 CAAG8002-0821 CAAG8002-0822 CAAG8002-0823 CAAG8002-0824 CAAG8002-0825 CAAG8002-0826
Capillaries for Thermo/Dionex LC Systems, Agilent Technologies
Tubing Chromatography Tubing
Stainless steel capillaries and tubing for selected Thermo Fisher Scientific HPLC systems.
  • Made of ultra-clean stainless steel tubing
  • Ideal for most standard applications, and the best choice for reliable high-pressure sealing
  • Smooth inner surface for the lowest backpressure
  • Precision cutting for burr-free, square-cut ends without inner-diameter distortion
  • Both ends accept standard fittings
  • Predefined lengths for specific path locations to avoid peak broadening
  • Individually tested to ensure reproducibility
  • Available in non pre-swaged
Agilent offers aMore Product Information