Dissolution Conversion Kits, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
12-6100 12-6368 27-6105
CAAG12-6100EA 1079.67 CAD
CAAG12-6100 CAAG12-6368 CAAG27-6105
Dissolution Conversion Kits, Agilent Technologies
Dissolution Apparatus
Agilent can also support small-volume dissolution with the 708-DS dissolution apparatus.
  • Special conversion kits: available in sets of 6 and 8, also available for the Chinese Pharmacopeia (CP) 250 ml vessel
  • Conversion kits are available to upgrade your 705-DS and 7000 and 7010 models to TruCenter vessels
The Agilent 708-DS dissolution apparatus can also support small-volume dissolution. Each position will require a conversion kit to hold a 100 or 200 ml vessel in position. The conversion kits are designed to be used with either mini paddles, miniMore Product Information