CTC Smart Autosampler Syringes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8010-1303 8010-1304 8010-1306 8010-1316 8010-1319 8010-1321 8010-1325 8010-1326 8010-1336 8010-1338 8010-1339 8010-1340 8010-1343 8010-1346 8010-1352
CAAG8010-1303EA 331.24 CAD
CAAG8010-1303 CAAG8010-1304 CAAG8010-1306 CAAG8010-1316 CAAG8010-1319 CAAG8010-1321 CAAG8010-1324 CAAG8010-1325 CAAG8010-1326 CAAG8010-1336 CAAG8010-1338 CAAG8010-1339 CAAG8010-1340 CAAG8010-1343 CAAG8010-1346 CAAG8010-1352
CTC Smart Autosampler Syringes, Agilent Technologies
Syringes Syringes for Chromatography
Agilent now offers Smart Syringes compatible with your PAL3 Autosampler!
  • Smart technology enables critical parameter tracking for each unique syringe
  • Compatible with all PAL3 models
  • CTC syringes provide precise and effective sample delivery from your CTC autosampler to Agilent GC
  • Handles both liquid and headspace injections
Agilent now offers smart syringes compatible with your PAL3 Autosampler! Each smart syringe is equipped with its own read/write chip that allow you to track critical parameters, such as the number of injections a specific syringe has made, the max temperature it’s experienced, and more! Furthermore, these smart syringes allow your PAL3 instrument to automatically recognize and upload these parameters, for efficient and detailed tracking. For precise, effective, sample delivery from your CTC autosampler, Agilent has the right syringe for you. Whether you're performing liquid or headspace injections on your PAL3 autosampler, you'll find what you need among our wide range of syringes for CTC autosamplers.