Interface Cones and Supplies for PerkinElmer ICP-MS Instruments, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8003-0576 8003-0580 8003-0583 8003-0584
CAAG8003-0576EA 3995.53 CAD
CAAG8003-0576 CAAG8003-0580 CAAG8003-0583 CAAG8003-0584
Interface Cones and Supplies for PerkinElmer ICP-MS Instruments, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Sampler, skimmer, hyper skimmer cones, & supplies for PerkinElmer NexION & ELAN ICP-MS instruments.
  • Guaranteed to be compatible with PerkinElmer ICP-MS instruments and qualified on a genuine PerkinElmer system
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent sampler, skimmer, hyper skimmer cones and supplies are equivalent to the PerkinElmer ICP-MS versions. You can have complete confidence in the compatibility of these parts with your PerkinElmer ICP-MS. Large-orifice sampler and skimmer cones provide superior long-term stability and resist clogging, allowing analysis under both high and low sample-uptake conditions.;Achieve guaranteed performance with interface cones and their supplies for PerkinElmer NexION 300/350/1000/2000 and ELAN 9000/6x00/DRC instruments from Agilent
Nickel interface cones provide the most economical operation and are recommended for the majority of sample types. Platinum interface cones provide better resistance to chemical attack and are recommended for more corrosive samples. Large-orifice sampler and skimmer cones provide superior long-term stability and resist clogging, allowing analysis under both high and low sample-uptake conditions.