Injectors and Injector Support Adaptors for PerkinElmer ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8003-0532 8003-0533 8003-0534 8003-0545 8003-0546 8003-0551
CAAG8003-0532EA 512.89 CAD
CAAG8003-0532 CAAG8003-0533 CAAG8003-0534 CAAG8003-0545 CAAG8003-0546 CAAG8003-0551
Injectors and Injector Support Adaptors for PerkinElmer ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Torch injectors and support adapters for PerkinElmer NexION 300/350 & ELAN 9000/6x00/DRCs ICP-MS.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent quartz, sapphire, and alumina torch injectors, and injector support adapters for PerkinElmer NexION 300/350 & ELAN 9000/6x00/DRCs ICP-MS instruments. Achieve guaranteed performance with supplies for PE from Agilent that are equivalent to the PerkinElmer ICP-MS versions so you can use supplies with complete confidence.
Torch injectors are available in a wide range of types. Quartz injectors are recommended for volatile organic solvents and a wide range of non-HF applications. Sapphire injectors are recommended for environmental, geochemistry, and HF applications.