Inert PFA Connector Tubes for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G3285-80023 G3285-80024 G4912-80016
CAAGG328580023KT 811.37 CAD
CAAGG328580023 CAAGG328580024 CAAGG491280016
Inert PFA Connector Tubes for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
PFA connecting tube, optional gas port, for the inert PFA sample introduction kit for Agilent ICP-MS.
  • PFA connecting tube optionally features a gas port enabling addition of option gas, O₂, for organics analysis
  • Connecting tube uses a push fit, O-ring free seal;Provides a perfectly smooth aerosol flow path eliminate droplet formation and improve washout
  • Manufactured from the highest purity grade PFA resin for the lowest blank contribution, ensuring it is suitable for semiconductor applications
The inert PFA sample introduction kit for Agilent ICP-MS instruments includes a PFA connecting tube joining the PFA spray chamber to the semi-demountable quartz torch. The connector optionally includes a gas port enabling addition of O₂ for organics analysis, or dilution gas for HMI applications. The connector fitting uses a push fit, O-ring free seal and has an ID that matches precisely to the ID of the spray chamber and torch injector holder on the exit side. This ensures a perfectly smooth aerosol flow path (no ledges or steps) helping to eliminate droplet formation and improving washout.