Graphite Tubes and Platforms for PerkinElmer GFAAS Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8003-0410 8003-0416 8003-0421 8003-0954 8003-0958
CAAG8003-0410PK 2951.37 CAD
CAAG8003-0410 CAAG8003-0416 CAAG8003-0421 CAAG8003-0954 CAAG8003-0958 CAAG8003-0959
Graphite Tubes and Platforms for PerkinElmer GFAAS Systems, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent graphite tubes and L'vov platforms for PerkinElmer THGA and HGA graphite furnace AA systems.
  • Guaranteed to be compatible with PerkinElmer Graphite Furnace AAS systems
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Instruction guides and how-to videos are available to ensure you get the best performance
More Product Information