Cleaning and Maintenance Supplies for GC/MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G1099-20030 G7077-60566 G1960-67100 3162-0110 5183-2096 5184-3571
CAG1099-20030EA 424.14 CAD
CAG1099-20030 CAAGG707760566 CAG1960-67100 CAAG3162-0110 CAAG5183-2096 CAAG5184-3571
Cleaning and Maintenance Supplies for GC/MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
A selection of replacement parts and cleaning supplies to keep your GC/MS running smoothly.
  • Ball, nut, and torx drivers available
  • Maintenance and source cleaning kits, and expendable supplies kits for systems using the DS102 rough pump
  • Cleaning powder for ion transfer capillary cleaning
Agilent offers a selection of replacement parts and cleaning supplies to help you keep your GC/MS running smoothly. Our range includes ion transfer capillary cleaning powder, ferrule kits, sorbent trap refills, and vacuum grease, among other products. Excessive background in the mass spectra, which can come from the GC or MS, indicates that the system requires maintenance. The source of contamination can sometimes be determined by identifying the contaminants. Some contaminants are much more likely to originate in the GC, while others are likely to originate in the MS.