Cells, Windows and Source lamps for PerkinElmer FIMS/FIAS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8003-0967 8003-0970 8003-0971
CAAG8003-0967EA 3599.66 CAD
CAAG8003-0967 CAAG8003-0970 CAAG8003-0971
Cells, Windows and Source lamps for PerkinElmer FIMS/FIAS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent FIAS/FIMS Cells, Windows and Source Lamps for PerkinElmer FIMS/FIAS systems.
  • Guaranteed to be compatible with PerkinElmer FIMS/FIAS systems
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent FIAS cell, FIMS cell, FIMS mercury source lamp and FIAS/FIMS cell replacement parts for PerkinElmer FIMS/FIAS systems are equivalent to genuine FIAS cell, FIMS cell, mercury source lamp and FIAS/FIMS cell replacement parts for PerkinElmer FIMS/FIAS systems. You, can be confident that these supplies will integrate seamlessly with your PerkinElmer instrument.