Burners and Supplies for PerkinElmer AAS Instruments, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8003-0395 8003-0418
CAAG8003-0395EA 355.59 CAD
CAAG8003-0395 CAAG8003-0418 CAAG8003-0858
Burners and Supplies for PerkinElmer AAS Instruments, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent burner heads for PerkinElmer PinAAcle/AAnalyst flame atomic absorption systems.
  • Burner heads are designed to provide the best precision, efficient drainage, minimal burner blockage, and reduced interferences
  • Guaranteed to be compatible with PinAAcle/AAnalyst flame atomic absorption systems
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent burner heads for PerkinElmer PinAAcle/AAnalyst flame AA systems are designed to provide the best precision, efficient drainage, minimal burner blockage, and reduced interferences. The 5 cm nitrous oxide burner head is required for nitrous oxide-acetylene operation, but can also be used with air-acetylene or rotated to provide reduced sensitivity. A 5 cm air-acetylene burner head is available for applications in which reduced sensitivity is required. It can be rotated to provide further sensitivity reduction, and has a wide slot to prevent clogging. The long pathlength 10 cm, single-slot air-acetylene burner head provides the best sensitivity for air-acetylene elements.