Application Kits for ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G8010-68005 G8010-68007 G8010-68008 G8010-68029
CAAGG801068005KT 6840.36 CAD
CAAGG801068005 CAAGG801068007 CAAGG801068008 CAAGG801068029
Application Kits for ICP-OES, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Convienent ICP-OES application kits containing matrix compatible sample introduction supplies.
  • Convenient application kits ensure you have compatible supplies for your application
  • Purchasing the kit provides a 15% savings over purchasing the parts individually
  • Inert applications kit provides inert sample introduction supplies to be used with samples containing free HF
  • Semi-volatile organic kit tobe used for common organic solvents like kerosene, xylene, and white spirit
  • Volatile organic application kits for use with volatile solvents such as gasoline
  • Ketone applications kit for use with ketone based solvents including MIBK
Agilent provides a large selection of convenient ICP-OES application kits to ensure that everything is available to run the most challenging samples. Application kits for ketone based organic samples, semi-volatile organic samples, volatile samples, and inert applications where HF is used are all offered. The application kits come with the correct torch, spray chamber, nebulizer, and peristaltic pump tubing required to run your application.