Application Kits for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G3280-60580 G4912-68000 G4912-68001 G4912-68002 G4912-68003
CAAGG328060580KT 5133.32 CAD
CAAGG328060580 CAAGG491268000 CAAGG491268001 CAAGG491268002 CAAGG491268003
Application Kits for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Convenient ICP-MS application kits include all sample introduction supplies required for success.
  • Convenient application kits ensure you have compatible supplies for your application
  • Save as much as 15% over the cost of purchasing the supplies and components individually
  • Organic kits provides components to enable analysis of common organic solvents like kerosene, and xylene
  • Semi-conductor application kits include high purity PFA sample introduction kits for low background and resistance to process chemicals
  • Inert application kits provides inert sample introduction supplies to be used with samples containing free HF
  • Speciation kits for environmental applications provide the components to interface HPLC systems
Agilent provides a large selection of convenient ICP-MS application kits to ensure that everything is available to run the most challenging samples. Application kits for organic solvents, semi-conductor samples, inert applications where HF is used, and speciation kits for environmental applications are all offered. The application kits include the correct components required to run your application, such as torch, spray chamber, nebulizer, peristaltic pump tubing or interfacing supplies. The application kits also provide a lower cost than purchasing the supplies individually.