Sample Loops for PerkinElmer FIMS/FIAS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8003-0679 8003-0681 8003-0680
CAAG8003-0679EA 201.94 CAD
CAAG8003-0679 CAAG8003-0681 AG8003-0680
Sample Loops for PerkinElmer FIMS/FIAS, Agilent Technologies
Chromatography Supplies Chromatography Sample Loops
Agilent sample loops are fully compatible with your PerkinElmer FIMS/FIAS systems, so can be confident that they will integrate seamlessly with your PerkinElmer system.
  • Guaranteed to be compatible with PerkinElmer FIMS/FIAS systems
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Worldwide technical support team to answer questions regarding part fit, function, or application-specific questions and concerns to help you get the most out of your PerkinElmer system
  • In stock and available worldwide for 24- to 48-hour delivery (in most regions)
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact