VGA 77 Supplies, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
9910040000 3710026390 6610011600 6910012600 7010017200 9910039900 9910040200 9910061800 9910062100 9910071100 9910093300 9910102500
CAAG9910040000PK 795.49 CAD
CAAG9910040000 CAAG3710026390 CAAG6610011600 CAAG6910012600 CAAG7010017200 CAAG9910039900 CAAG9910040200 CAAG9910061800 CAAG9910062100 CAAG9910071100 CAAG9910093300 CAAG9910102500
VGA 77 Supplies, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Typical supplies for VGA 77 Vapor Generation Accessory used with Agilent flame AA Spectrometers.
  • The T-shaped open ended quartz absorption cell is used with the VGA 77 when determining hydride forming elements
  • The small internal volume provides more uniform heating and more efficient atomization
  • The closed, flow through cell provides better precision and sensitivity for cold vapor mercury determinations, and the end-mounted entry and exit ports facilitate collection of mercury vapors to comply with local discharge laws
  • The gas-liquid separator for the VGA 77 facilitates the release of gas from the liquid mix, while inhibiting the formation of large bubbles. The small volume above the liquid helps achieve steady state conditions quickly
  • The hydride module for the VGA-77 incorporates the plumbing manifold and the gas-liquid separator, and is interchangeable
  • A separate hydride module should be used exclusively for Hg determinations to reduce cross contamination
  • The hydride module is easily removed to enable change-over between determinations of hydride forming elements and mercury. It also incorporates space to hold the sample and reagent bottles
  • Generation of the volatile metal hydrides use sodium borohydride as a reducing agent
The Agilent fully automated modular VGA 77 vapor generation accessory is used with Agilent flame AA systems to. enables rapid trace level determinations of mercury and other hydride-forming elements at parts-per-billion (μg/L) concentrations using the vapor or hydride generation technique. The VGA 77 offers the convenience of flame AA determinations, unmatched precision at ppb levels, and ease-of-use from the continuous flow operation.
The entire plumbing assembly is integrated into a separate hydride module, which can be simply exchanged when switching between elements. This reduces change-over times and eliminates cross-contamination from by the conflicting chemistries required for each element.