Valve Replacement Parts for Thermo/Dionex LC Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8002-0602 8002-0807 8002-0860
CAAG8002-0602EA 253.76 CAD
CAAG8002-0602 CAAG8002-0603 CAAG8002-0807 CAAG8002-0812 CAAG8002-0860
Valve Replacement Parts for Thermo/Dionex LC Systems, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
A comprehensive range of replacement valve parts for selected Thermo Fisher Scientific HPLC systems.
  • Vespel is a polyimide with low wear and high chemical resistance recommended for use with solutions under pH 10
  • More basic solutions dissolve Vespel over time and damage the rotor seal
  • Tefzel is recommended for use in applications such as higher concentrations of methylene chloride or dimethyl sulfoxide (DMSO)
Agilent valve supplies work seamlessly with selected Thermo Fisher Scientific HPLC systems for reliable, accurate, and reproducible results.More Product Information